Learn More
PIP™ Bouton™ Captain™ Rimless Safety Glasses
Click to view available options
Frame Color:
Clear
Gray
Lens Coating:
Anti-fog, Scratch-resistant
Anti-reflective, Scratch-resistant
Scratch-resistant
Lens Tint:
Blue, Indoor/Outdoor
Clear
Gray
Description
- Lightweight, rimless design provides all day user comfort
- Bayonet temples with end tips that curve out for ease of placement on face
- Non-slip rubber nose pads offer a secure fit
- Clear Lens for clear, crisp and distortion free visibility while meeting the requirement standards that help provide protection from projectiles and impact in general use applications
- Treated front and back with a clear, scratch-resistant coating for a much harder surface that is more resistant to scratching
- Anti-Fog Coating eliminates the condensation of moisture on lenses that causes fogging so your lenses and vision stay clear
Specifications
Lens Coating | Anti-fog, Scratch-resistant |
Lens Tint | Clear |
Product Type | Safety glasses |
Frame Color | Clear |
Lens Material | Polycarbonate |