Learn More
Hach Company Bottle, BOD, 300 mL, 6/pk
Supplier: Hach Company 62106
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The standard for BOD analysis. Borosilicate glass bottle with flared lip and ground-glass stopper to prevent air entrapment and for water seal. Frosted marking area. Stopper included (60 and 300 mL sizes). Serialized bottles have silk-screened, sequential