Learn More
Wilmad Labglass BOTTLE SPECIFIC GRAVITY 50ML
Supplier: Wilmad Labglass LG3530108
Bottle, Specific Gravity, Gay-Lussac, Unadjusted. Capacity: 50mL; Joint Size: [ST] 10/18. Unadjusted specific gravity bottle. With standard taper capillary stopper.