Learn More
Wilmad Labglass BOTTLE SPECIFIC GRAVITY 2ML
Supplier: Wilmad Labglass LG3530100
Bottle, Specific Gravity, Gay-Lussac, Unadjusted. Capacity: 2mL; Joint Size: [ST] 7/15. Unadjusted specific gravity bottle. With standard taper capillary stopper.