Learn More
Wilmad Labglass BOTTLE SPECIFIC GRAV 5ML ADJ
Supplier: Wilmad Labglass LG3540102
Bottle, Specific Gravity, Gay-Lussac, Adjusted. Capacity: 5mL; Joint Size: [ST] 7/15. Adjusted at 20degreeC. The capillary stoppers are not interchangeable since the final adjustment for volume is made on the stopper.