Learn More
Wilmad Labglass BOTTLE SPECIFIC GRAV 50ML ADJ
Supplier: Wilmad Labglass LG3540108
Bottle, Specific Gravity, Gay-Lussac, Adjusted. Capacity: 50mL; Joint Size: [ST] 10/18. Adjusted at 20degreeC. The capillary stoppers are not interchangeable since the final adjustment for volume is made on the stopper.