Learn More
Wilmad Labglass BOTTLE SPECIFIC GRAV 2ML ADJ
Supplier: Wilmad Labglass LG3540100
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Bottle, Specific Gravity, Gay-Lussac, Adjusted. Capacity: 2mL; Joint Size: [ST] 7/15. Adjusted at 20degreeC. The capillary stoppers are not interchangeable since the final adjustment for volume is made on the stopper.
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov