Learn More
Wilmad Labglass BOTTLE SPC GRAV PYCNOMTR 50ML
Supplier: Wilmad Labglass LG3550104
Bottle, Specific Gravity, Pycnometer, Complete. Capacity: 50mL. With a conical flask body to give maximum stability. Supplied with flask, thermometer, and flask side arm cap.