Learn More
Wilmad Labglass BOTTLE SPC GRAV PYCNOMTR 25ML
Supplier: Wilmad Labglass LG3550102
Bottle, Specific Gravity, Pycnometer, Complete. Capacity: 25mL. With a conical flask body to give maximum stability. Supplied with flask, thermometer, and flask side arm cap.