Learn More
Wilmad Labglass BOTTLE SPC GRAV PYCNOMTR 100ML
Supplier:  Wilmad Labglass LG3550106
Encompass_Preferred
Bottle, Specific Gravity, Pycnometer, Complete. Capacity: 100mL. With a conical flask body to give maximum stability. Supplied with flask, thermometer, and flask side arm cap.
Catalog No. 50-806-160
Item Discontinued This item has been discontinued by Fisher Scientific. Please to view product availability in your area.