Learn More
Wilmad Labglass BOTTLE SPC GRAV HUBB-CARM 25ML
Supplier: Wilmad Labglass LG3542100
Bottle, Specific Gravity, Hubbard-Carmick. Capacity: 25mL. Used in standard test methods for specific gravity of viscous fluids, semi-solid bitumens and emulsions.