Learn More
Med Vet International Plastic Boston Round Wash Bottle, 150ml, each
Supplier: Med Vet International PWB150
Wash bottle used to wash labware and to store washing solutions, such as acetone or deionized waterLDPE body for flexibility, elasticity, and impact, stress-cracking, and moisture resistance, and is lightweight. Boston round shape for applications that require a rounded body, short neck, and narrow mouth to store liquid samples and solutions, and dry materials. Polypropylene screw closure with its angled stem and draw tube molded in one piece for leak-proof applications. Spout can be cut back to increase the flow of washing solution.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.