Learn More
Hach Company Disposable BOD bottle, 300 mL
Supplier: Hach Company 2943101
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Single 300 mL disposable BOD bottle without stopper. Plastic bottle is EPA approved for BOD measurement. Box of 100 bottles available under article number 2943100. Acrylic stoppers (25 pack) available under article number 2943900. Glass stoppers (quantity of 1) available under article number 190901.