Learn More
Chemglass Life Sciences 4,000mL Bottle, Aspirator, Graduated
Supplier: Chemglass Life Sciences CG112405
4,000mL Bottle, Aspirator, Graduated. Aspirator bottles with permanent white graduations. Useful as a reservoir for fermentation work or as a large scale animal feeder. Bottle has a reinforced tooled top to accept the listed size of rubber stopper. Hose connection has an O.D. of 9mm at the largest serration. Bottles are also available plastic coated.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.