Learn More
Ace Glass, Inc. Bottle, pressure, 8mL, 60psi, complete with #7 front-seal plug
Supplier: Ace Glass, Inc. 8648230
8mL pressure bottle with #7 Ace-Thred, rated for 60psig at room temperature. Includes #7 front-seal plug.