Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences Wheaton™ Boston Round Reagent Bottle, LDPE
Click to view available options
Capacity (Metric):
125 mL
Description
- Low Density Polyethylene, translucent color
- LDPE is very flexible with high impact strength
- Excellent for mild and strong buffers, good chemical resistance
- Narrow mouth ideal for liquids
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Capacity (Metric) | 125 mL |
Color | Natural |
Material | LDPE |
Quantity | 72/Case |
Closure Color | White |
Closure Included | Yes |
Mouth | Narrow |
Shape | Round |
Product Type | Reagent Bottle |
Closure Liner | Polyethylene Foam |
Show More |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction