Learn More
Eisco™ Borosilicate Glass Gooch Crucible
Gooch type crucible made of high quality Borosilicate 3.3 glass features a sintered disc with G-3 porosity
$14.00 - $16.75
Specifications
Description | Gooch Type |
---|---|
Material | Borosilicate Glass (3.3) |
Porosity | G-3 |
Description
- Made of high quality, heavy duty, Borosilicate 3.3 glass
- Features sintered disc with G-3 porosity
- Perfect for home, classroom & professional laboratories
Specifications
Gooch Type | |
Borosilicate Glass (3.3) | |
G-3 |