Learn More
Fine Science Tools Bone Scissors, Tungsten Carbide, Straight, 12.5 cm
SDP
Supplier:  Fine Science Tools 1603012
Encompass
Tip Shape: Straight/ Cutting Edge: 15mm/ Alloy / Material: Stainless Steel/ Length: 12.5cm/ Joint Action: Single/ Feature: Tungsten Carbide
Catalog No. NC0264351