Learn More
Electron Microscopy Sciences Bone Saw with rounded end, 203mm blade
Supplier: Electron Microscopy Sciences 7289520
Bone saw with a rounded end and 203 mm blade. The inside handle grip is 70 mm and the finger opening is 25 mm.