Learn More
Chemglass Life Sciences Bonded Closure, Black, 13-425, 0.60" PTFE/Silicone with Slit
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Chemglass Life Sciences CV38420013
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Bonded Closure, Black, 13-425, 0.60" PTFE/Silicone with Slit. Bonded closures feature clear PTFE/Silicone bonded directly to the closure without the use of glue or adhesives. These bonded closures provide a cost effective, integrated, one-piece system that eliminates costly line fallout. Pre-slit design.