Learn More
Chemglass Life Sciences Bonded Closure, Black, 13-425, 0.60" PTFE/Silicone
Supplier: Chemglass Life Sciences CV38400013
Bonded Closure, Black, 13-425, 0.60" PTFE/Silicone. Bonded closures feature clear PTFE/Silicone bonded directly to the closure without the use of glue or adhesives. These bonded closures provide a cost effective, integrated, one-piece system that eliminates costly line fallout. Non-slit designs.