Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Body Scientific International™ Post-it™ Disease 20 Chart Series Chart
SCI_ED

A simplified teaching and learning tool showing 20 diseases in visual format with illustrations; perfect for students and patients.

Supplier:  Fischer Technical Company BS401

Catalog No. S02586

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Description

Description

 

  • Feature detailed drawings and identifications
  • Printed on heavy, quality stock paper, backed with 3M adhesive Post-It material
  • Stick and re-stick to virtually any surface
  • Can be reposition without damaging the surface

 

Componants of series:
FLU Prevention
Influenza A
Osteoarthritis
Rheumatoid Arthritis
Emphysema
Bronchitis
Asthma
GERD
Atherosclerosis
Complication Of Athero
Hypertension
Stroke
OtitisMedia
Acute Coronary Disease
Breast Cancer
Complications Of Diabetes
Type 2 Diabetes
Overactive Bladder (female)
BPH
Osteoporosis

Order Info

Dimension: 11W x 17 in. L

Specifications

Specifications

Disease 20 Chart Series
Science
17 in.
11 in.
Flu prevention, influenza a, osteoarthritis, rheumatoid arthritis, emphysema, bronchitis, asthma, gerd, atherosclerosis, complication of athero, hypertension, stroke, otitismedia, acute coronary disease, breast cancer, complications of diabetes, type 2 diabetes, overactive bladder (female), bph, osteoporosis
Paper
17 x 11 in. (43.18 x 27.84 cm)
43.18 cm
27.84 cm
Simplified teaching and learning of osteoarthritis in visual format with illustration for students and patients
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit