Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Body Scientific International™ Post-it™ Anatomy of Respiratory System Chart
A perfect teaching and learning aid with illistration to convey all the primary points on Respiratory system.
Supplier: Fischer Technical Company BS303
Description
- Feature detailed drawings and identifications
- Printed on heavy, quality stock paper, backed with 3M adhesive Post-It material
- Stick and re-stick to virtually any surface
- Can be reposition without damaging the surface
Order Info
Dimension: 11W x 17 in. L
Specifications
Anatomy of Respiratory System Chart | |
Science | |
17 in. | |
11 in. | |
Perfect teaching and learning aid with illustration to convey all the primary points on respiratory system |
Paper | |
17 x 11 in. (43.18 x 27.84 cm) | |
43.18 cm | |
27.84 cm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction