Learn More
Enzo Life Sciences Bone morphogenetic protein 4 (human), (recombinant) (10µg)
Supplier: Enzo Life Sciences ENZPRT2010010
Alternative name: BMP-4. Purity: ≥95% (SDS-PAGE). Formulation: Lyophilized from 0.7 mg/mL in 2X PBS containing 6% ethanol. Source: Produced in HEK cells. Glycosylated disulfide linked homodimer. Long Term Storage: -20°C.