Learn More
Electron Microscopy Sciences Blue Silicone Rubber Kit 1 GL
Supplier: Electron Microscopy Sciences 24230B
Blue Silicone Rubber is extremely high tear, high tensile strength, two components, and tin catalyzed RTV silicone rubber. It is designed for casting polyurethane foam, polyester, and epoxy parts. It is a superior product with the following advantages over the RTV silicones.
Properties include extremely high tear strength, low shrinkage, low viscosity, excellent chemical resistance, and excellent shelf aging. The catalyst is color codes. IT also has a long pot life, is nsensitive to inhibition, a variable cure rate, and good dielectric properties.
Blue Silicone Rubber comes as a kit consisting of 1 gal (9 lb) silicone base and 1 pt (1lb) blue catalyst.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.