Learn More
Andwin Scientific BLUE LITE AUTORAD FILM
Supplier: Andwin Scientific F90248X10
The 8" x 10" GeneMate blue autoradiography film is ideal for different applications like the chemiluminescence and autoradiography of 32P and 125I. This product lasts 2 years from the date of the manufacturing. It is sold in a box of 100.