Learn More
Medicus Health Hardside Cooler - Compact 7.5 Qt
Supplier: Medicus Health 5323M1
∙ Compact 7.5 quart medical cooler is perfect for transporting specimens to the lab and maintaining temperatures.∙ Great option for Blood Banks to send units to the ER or Surgery to help prevent units from returning over temperature∙ Hard side blood transport coolers have feet and can be stacked for storage∙ Stainless Steel hardware on this cooler for medical transport will provide many years of durable service∙ Convenient handle and heavy duty shoulder strap ∙ Constructed of durable polypropylene for all types of specimen transport needs∙ Insulated with high-grade molded polystyrene foamnon-absorbent surface ∙ Easy to clean and disinfect∙ Airtight closure seal is provided by an EVA Gasket Seal recessed into the lid∙ Internal Dimensions: 10.5"W x 7.25"D x 6.25"H; External Dimensions: 12.25"W x 8"D x 10"H
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.