Learn More
Serim Research Corp SERIM GUARDIAN™ Blood Leak Test Strips
Supplier: Serim Research Corp 5119C
SERIM Blood Leak Test Strips provide a rapid method to test dialysate for blood if a dialyzer membrane leak is suspected during the hemodialysis procedure. The test can assist in differentiating an actual blood leak from a false alarm. Kit contains 2 bottles of 25 tests