Learn More
Medicus Health Blood Bank Transport Cooler, 13 Quart
SDP
Supplier:  Medicus Health 5318M5
Encompass
∙ 13 Quart Blood Bank Transport Cooler can hold up to 6 Units of blood or other products and three ice packs ∙Includes a prominent sticker to remind staff to return units to the Blood Bank∙Great option for Blood Banks to send units using this blood cooler to the ER or Surgery to help prevent units from returning over temperature∙Stainless Steel hardware will provide many years of durable service∙Convenient handle on top and heavy duty strap on this medical transport cooler can be used to secure lid during transport or be used as a shoulder strap∙Constructed of durable polypropylene for easy cleaning∙Insulated with High-Grade molded Polystyrene Foam∙Non-absorbent surface that's easy to clean ∙Offers superior protection for your blood products∙Airtight closure seal is provided by an EVA Gasket Seal recessed into the lid∙Internal Dimensions: 12"L x 7"W x 8.5"H; External Dimensions: 15"L x 10"W x 11"H
Catalog No. NC1489359