Learn More
Thermo Scientific™ Blood Agar (TSA with 5% Sheep Blood) / MacConkey Agar Plate
Grow fastidious organisms, detect and enumerate coliforms, and isolate and detect Salmonella and Shigella spp. on a single plate.
$109.00 - $1110.00
Specifications
Color | Opaque red/ clear pink |
---|---|
Content And Storage | 2°C to 8°C |
Description | Blood Agar (TSA with 5% Sheep Blood) / MacConkey Agar Plate |
Format | 85mm biplate |
Manufacturing Origin | United States |
Catalog Number | Mfr. No. | Description | dimFormat | Quantity | Manufacturing Origin | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | dimFormat | Quantity | Manufacturing Origin | Price | Quantity & Availability | ||
R02049
|
Thermo Scientific™
R02049 |
Blood Agar (TSA with 5% Sheep Blood) / MacConkey Agar Plate | 100/Pk. | United States |
Case of 100 for $1,110.00
|
|
|||
R02050
|
Thermo Scientific™
R02050 |
Blood Agar (TSA with 5% Sheep Blood) / MacConkey Agar Plate | 10/Pk. | United States |
Pack of 10 for $109.00
|
|
|||
Description
Isolate and cultivate fastidious microorganisms with clearly visible hemolytic reactions (staphylococci and streptococci) and detect and enumerate coliform organisms such as E. coli, while at the same time isolating and detecting Salmonella and Shigella spp., using Thermo Scientific™ Blood Agar (TSA with 5% Sheep Blood) / MacConkey Agar.
Read more results from a single patient sample and produce more diagnostic details of pathogenic organisms while reducing storage capacity issues in the incubator with a bi-plate that includes two widely used microbiological media. One half of the plate contains TSA with 5% sheep blood to promote growth of fastidious organisms with clearly visible hemolysis forms for staphylococci and streptococci. MacConkey Agar on the other half of the plate is a selective medium for differentiating between coliforms and lactose and non-lactose fermenters.Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Opaque red/ clear pink | |
Blood Agar (TSA with 5% Sheep Blood) / MacConkey Agar Plate | |
United States | |
10 mL |
2°C to 8°C | |
85mm biplate | |
Agar | |
Flow Wrapped - Single Layer |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.