Learn More
Thermo Scientific™ Block, 6 x 1.5 mL, EMD

For Use With: Thermo Scientific™ Compact Dry Bath/Block Heater
Supplier: Thermo Scientific™ 88871103/EMD
Description
Increase your application capabilities with a variety of accessories for the Thermo Scientific™ Compact Dry Baths/Block Heaters.
Warranty: 2 yearsSpecifications
6 x 1.5mL block | |
6 | |
36 x 54 x 30 mm (1.4 x 2.1 x 1.2 in.) |
Thermo Scientific™ Compact Dry Bath/Block Heater | |
Accessory, Block |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.