Learn More
Waters Corp Black, 15 mm Screw Neck Cap and Preslit PTFE/Silicone Septum, 1000/pk
Supplier: Waters Corp 186001919
This septum is electron bonded to the cap and will not come dislodged during shipment or use.