Learn More
Waters Corp Black, Screw Neck Cap and Preslit PTFE/Silicone Septum, 100/pk
Supplier: Waters Corp 186000842
This septum is electron bonded to the cap and will not come dislodged during shipment or use.