Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Wells Lamont™ Black/Hi Vis Performance Quarter Socks
Click to view available options
Size: Large
Description
- Men's Performance Work Socks provide support and protection for your feet while you work.
- The mesh upper foot is designed for ventilation and moisture evaporation, keeping your feet dry throughout the day.
- Socks feature reinforced heel and toe as well as an Achilles heel pad for added protection and comfort.
- The socks non-binding top do not slip down and provide all-day gentle comfort.
- Added compression at the arch provides added support and stability.
Specifications
Size | Large |
For Use With (Application) | Cleanroom Use |
Material | Polyester, Nylon, Spandex |
Color | Hi-vis Black |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction