Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher Science Education™ Biuret Reagent
For protein test
$2.75 - $12.25
Specifications
Chemical Name or Material | Biuret Reagent |
---|---|
Color | Blue |
CAS | 7759-99-8, 7681-11-0, 6381-59-5, 131-73-2, 7732-18-5 |
Assay Percent Range | <1% w/v |
Packaging | Solution |
Description
For protein testSpecifications
Biuret Reagent | |
7759-99-8, 7681-11-0, 6381-59-5, 131-73-2, 7732-18-5 | |
Solution |
Blue | |
<1% w/v | |
Liquid |
Safety and Handling
ShelfLife : 2 years
DOTInformation : DOT Class 8, III, : Corrosive
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title