Learn More
Bismuth Lead eutectic wire, 0.79mm (0.031in) dia, Thermo Scientific Chemicals
$311.00 - $851.65
Specifications
Chemical Name or Material | Bismuth Lead eutectic wire |
---|---|
Name Note | 0.79mm (0.031 in.) dia. |
Melting Point | 124°C |
Avg. Mol. Wt. or Mol. Wt. Range | Bi:Pb; 55.5:44.5 wt% |
MDL Number | MFCD00801141 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AA44238Q4
|
Thermo Scientific Chemicals
044238Q4 |
0.25 lb. |
Each for $311.00
Order in Bulk
Request bulk or custom formats |
|
|||||
AA44238P2
|
Thermo Scientific Chemicals
044238P2 |
1 lb. |
Each
for $851.65
Save 16% Reg :
Order in Bulk
Request bulk or custom formats |
|
|||||
Description
Employed in chemical research. LBE is suggested as a coolant in some nuclear reactors and is a proposed coolant for the lead-cooled fast reactor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Bismuth Lead eutectic wire | |
124°C | |
MFCD00801141 | |
Wire |
0.79mm (0.031 in.) dia. | |
Bi:Pb; 55.5:44.5 wt% | |
Insoluble in water. |
Safety and Handling
GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.