Learn More
Bismuth Lead eutectic ingot, 99.9% (metals basis), Thermo Scientific Chemicals
Low melting application: fuses, etc
Supplier: Thermo Scientific Chemicals 040949P2
Description
Low melting application: fuses, etc
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Bismuth Lead eutectic ingot | |
Bi:Pb; 55.5:44.5 wt% | |
MFCD00801141 | |
99.9% |
124°C | |
(metals basis) | |
1 lb. | |
Ingot |
Safety and Handling
GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c
H302+H332-H350-H360Df-H373
TSCA : Yes
Recommended Storage : Ambient temperatures
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
RUO – Research Use Only