Learn More
Vector Laboratories Sambucus Nigra Lectin (SNA, EBL), Biotinylated, 2mg
Supplier: Vector Laboratories B13052
Sambucus nigra lectin, isolated from elderberry bark, binds preferentially to sialic acid attached to terminal galactose in α-2,6 and to a lesser degree, α-2,3 linkage. Binding is also inhibited to some extent by lactose or galactose. This lectin appears to bind sialic acid linked to N-acetylgalactosamine or galactose. SNA has been reported to inhibit cell-free protein synthesis. 2mg; Solution: 10 mM HEPES, 0.15 M NaCl, pH 7.5, 0.08% sodium azide, 0.1 mM CaCl2.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.