Learn More
Rockland Immunochemicals BIOTIN BETA GALACTOSIDASE Conjugated - B000-17
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Rockland Immunochemicals B00017
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Biotin is a small biomolecule important for many cellular processes. Most importantly for biotechnology applications, biotin is ammenable to conjugation to proteins for use in biochemical assays. Biotin has a very strong affinity for avidin and streptavidin; an attraction that is the strongest and most stable non-covalent interaction known.Beta-galactosidase (beta-gal; β-gal) is an enzyme that hydrolyzes β-galactosides (such as lactose and various glycoproteins) into simple sugars. Beta-galactosidase is routinely used as a reporter in biochemical experiments as well as a marker for gene expression experiments in E. coli. Beta-galactosidase can be used to cleave the chemical X-gal that forms an intense blue product that can easily be quantified.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.