Learn More
Agilent BioTek™ BioSpa 8 Automated Incubator, IVD PROMO
The BioTek™ BioSpa live cell analysis system offers unique benefits for a wide variety of live cell imaging applications in up to 8 microplates or other labware.
Supplier: Agilent BioTek™ BIOSPAGSI2358
This item is not returnable.
View return policy
Description
With 5 imaging modes, the BioTek BioSpa live cell analysis system enables a wide range of microscopy applications.
- Capture detailed cellular images, get cell-level analyses for migration and wound healing assays, 3D spheroid formation, and many other kinetic applications
- Built-in scheduling, environmental monitoring, and available liquid handling for walk away automation
- Multiple users can run simultaneous processes for maximum efficiency in the lab
- For In Vitro diagnostic use
Specifications
Ambient to 45°C | |
20.1 in. | |
18.9 in. | |
80 to 95% | |
27.2 in. | |
Automated Microplate Incubator |
CE and TUV Marked, RoHS Compliant | |
51 cm | |
48 cm | |
CO2/O2 Control and Monitoring, Water Pan and Humidity Monitoring, BioSpa Software, One Alignment Kit | |
69 cm | |
8 Plates |