Learn More
Abbott™ Biosite™ Triage™ Drugs of Abuse Panel
Immunoassay for the qualitative determination of the presence of parent compound and/or major metabolites of up to nine distinct drug classes in urine
Specifications
Clia Complexity | Moderate |
---|---|
Sample Type | Urine |
Includes | Amphetamines, Methamphetamines, Barbiturates, Benzodiazepines, Cocaine, Marijuana, Methadone, Opiates, PCP, Propoxyphene and Tricyclic Antidepressants |
Quantity | 25 Tests |
Test Time | 10 min. |
Catalog Number | Mfr. No. | Quantity | Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Type | Price | Quantity & Availability | ||||
23-034-973
|
Abbott™
92000 |
25 Tests | Triage™ Drugs of Abuse Panel plus TCA | Item Discontinued Due to product restrictions, please Sign In to purchase or view availability for this product.This item has been discontinued by the supplier. | N/A | ||||
Description
- Test results from a urine sample in about 10 minutes
- CLIA Complexity: Moderate
- A self-contained, disposable, credit card-sized test device
- Easy-to-read results for up to nine drug classes
- Tests include amphetamines/methamphetamines, barbiturates, benzodiazepines, cocaine, marijuana, methadone, opiates, PCP and tricyclic antidepressants
- The presence of color in any drug detection zone indicates the presence of the drug at or above the threshold concentrations
- Built-in positive and negative controls in every device validate results
- Kit sizes available in 25 test configurations
Specifications
Moderate | |
Amphetamines, Methamphetamines, Barbiturates, Benzodiazepines, Cocaine, Marijuana, Methadone, Opiates, PCP, Propoxyphene and Tricyclic Antidepressants | |
10 min. |
Urine | |
25 Tests |
Spot an opportunity for improvement?
missing translation for 'provideContentCorrection'
missing translation for 'provideContentCorrection'
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
missing translation for 'productTitle'