Learn More
Alere ExpressTest™ 5 Drug Panel
Immunoassay for the qualitative determination of the presence of parent compound and/or major metabolites of five distinct drug classes in urine
Supplier: Alere 93005/DEL
Includes: Reagents for 25 tests
Description
- Utilizes SAMHSA recommended screening cut-off concentrations
- 5-test panel detects marijuana (THC), opiates (OPI), phencyclidine (PCP), cocaine (COC), and amphetamines (AMP/mAMP)
- CLIA Complexity: Moderate
- Urine sample
- Presence of color in any drug detection zone indicates the drug is at or above the recommended SAMHSA cutoff
- Built-in positive and negative controls in every device validate results
- Reagents are stable at room temperature up to 20 months