Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Germaine™ Laboratories BioSign™ Flu A/B Kit
An In vitro rapid qualitative test that detects influenza type A and type B antigens directly from nasal swab, nasopharyngeal swab and nasal aspirate/wash specimens.
Supplier: Germaine™ Laboratories BSP51025P
This item is not returnable.
View return policy
Description
- Differentiates between Influenza Type A and B on a single test device
- Produces results in 10 to 15 minutes
- Each test device includes built-in controls
- Each kit comes with external controls
- Meets FDA performance criteria for influenza tests stated in code 21 CFR 866.3328
- CLIA Complexity: Moderate complexity when used with nasal wash/aspirate samples
- CLIA Waived when used with nasal and nasopharyngeal swabs
Specifications
2°C to 30°C | |
Waived (Nasal Swab) | |
Positive and Negative Control Swabs |
Yes | |
Influenza A and B | |
6 x 25 Kits = 150 Tests |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction