Learn More
MilliporeSigma™ Supelco™ BIOshell™ lgG, C4 HPLC Column, 1,000 Å, 2.7 μm Particle Size
BIOshell™ IgG 1,000 Å C4 is a high-speed, high-performance liquid chromatography column based on a new wide-pore (1, 000 Å) Fused-Core™ particle design.
$763.00 - $1522.00
Specifications
Particle Size | 2.7 μm |
---|---|
pH | 2 to 9 |
Pore Size | 1000 Å |
Surface Area | 22 m2/g |
Stationary Phase | C4 (Butyl) |
Catalog Number | Mfr. No. | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
11-100-7385
|
MilliporeSigma™ Supelco™
63281U |
2.1 mm | 2 cm |
Each for $787.00
|
|
||||
11-100-7386
|
MilliporeSigma™ Supelco™
63282U |
2.1 mm | 3 cm |
Each for $915.00
|
|
||||
11-100-7387
|
MilliporeSigma™ Supelco™
63283U |
2.1 mm | 5 cm |
Each for $888.00
|
|
||||
11-100-7195
|
MilliporeSigma™ Supelco™
63284U |
2.1 mm | 7.5 cm |
Each for $1,146.00
|
|
||||
11-100-6990
|
MilliporeSigma™ Supelco™
63288U |
2.1 mm | 10 cm |
Each for $1,376.00
|
|
||||
11-100-6755
|
MilliporeSigma™ Supelco™
63289U |
2.1 mm | 15 cm |
Each for $1,522.00
|
|
||||
11-100-7388
|
MilliporeSigma™ Supelco™
63306U |
3 mm | 2 cm |
Each for $763.00
|
|
||||
11-100-7389
|
MilliporeSigma™ Supelco™
63307U |
3 mm | 3 cm |
Each for $763.00
|
|
||||
11-100-7390
|
MilliporeSigma™ Supelco™
63308U |
3 mm | 5 cm |
Each for $888.00
|
|
||||
11-100-7196
|
MilliporeSigma™ Supelco™
63311U |
3 mm | 7.5 cm |
Each for $1,166.00
|
|
||||
11-100-6991
|
MilliporeSigma™ Supelco™
63313U |
3 mm | 10 cm |
Each for $1,293.00
|
|
||||
11-100-6756
|
MilliporeSigma™ Supelco™
63314U |
3 mm | 15 cm |
Each for $1,430.00
|
|
||||
11-100-7391
|
MilliporeSigma™ Supelco™
63322U |
4.6 mm | 2 cm |
Each for $763.00
|
|
||||
11-100-7392
|
MilliporeSigma™ Supelco™
63324U |
4.6 mm | 3 cm |
Each for $763.00
|
|
||||
11-100-7197
|
MilliporeSigma™ Supelco™
63327U |
4.6 mm | 7.5 cm |
Each for $1,166.00
|
|
||||
11-100-7393
|
MilliporeSigma™ Supelco™
63325U |
4.6 mm | 5 cm |
Each for $888.00
|
|
||||
11-100-6992
|
MilliporeSigma™ Supelco™
63328U |
4.6 mm | 10 cm |
Each for $1,293.00
|
|
||||
11-100-6757
|
MilliporeSigma™ Supelco™
63329U |
4.6 mm | 15 cm |
Each for $1,477.00
|
|
||||
Description
BIOshell™ lgG, C4 columns contain fused-Core particles that provide a thin porous shell of high-purity silica surrounding a solid silica core. This particle design exhibits very high column efficiency due to the shallow diffusion paths in the 0.5-micron thin porous shell and the small overall particle size of 2.7-microns. The densely bonded, extensively endcapped dimethylbutyl stationary phase of BIOshell™ IgG 1,000Å C4 provides a stable, reversed-phase packing that can be used for separating high molecular weight compounds such as proteins.Specifications
2.7 μm | |
1000 Å | |
C4 (Butyl) | |
C4 (Butyl) Phase/Fused-Core Particle Platform | |
Reversed Phase | |
Fused-Core HPLC |
2 to 9 | |
22 m2/g | |
BIOshell | |
Yes | |
Stainless Steel Hardware |
BIOshell is a trademark of Sigma-Aldrich Co. LLC. Fused-Core is a registered trademark of Advanced Materials Technology, Inc.