Learn More
Medex Supply BIOPSY PUNCH 6 MM 25/BOX
SDP
Supplier:  Medex Supply HLBP60
Encompass_Preferred
Description of the Health Link Biopsy Punch, Sterile 6.0 mm Seamless sharp blade. Hollow handle. Safety capped. Sterile, single use disposable. Sterility indicator dot gives visual evidence of sterility. Tamper evident seal. Gamma Sterilized (cell and tissue cultures suffer adverse effects or sometimes are destroyed when exposed to EtO residues). Sure Grip handle. Color coded handle embossed with size. Easy to peel "no fiber tear" pouch. Dispenser Box: 25 individual pouches per box. Biopsy Punch now has an expiration date and the shelf life is 12 months.
Catalog No. NC9543097