Learn More
Aniara Diagnostica LLC BIOPHEN FVIII:C ASSAY
Supplier:  Aniara Diagnostica LLC A221402RUO
Encompass_Preferred
This item MFG by HYPHEN BioMed and Dis. by Aniara Diagnostics. BIOPHEN Factor VIII:C Assay Kit. Quantitative chromogenic assay for Factor VIII:C activity in human citrated plasma, biological fluids, or FVIII concentrates. Manual Microplate (2x50 Tests) or Automated.
Catalog No. 50-100-8885
Explore available promotions