Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LaMotte™ BioPaddles™ Dual-Agar Paddles for Environmental Education
Contain microbe-specific media enclosed in a sterile vial.
$60.50
Specifications
Content And Storage | 18°C to 25°C |
---|---|
Manufacturing Origin | United States |
Sterility | Sterile |
Includes | Free basic microbe identification application and access to technical information sheet |
Packaging Type | Paddle |
Includes: Free basic microbe identification app and access to technical information sheet
Catalog Number | Mfr. No. | For Use With (Application) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Application) | Price | Quantity & Availability | |||||
S04344
|
LaMotte™
5550 |
Environmental education: cultivation of a wide variety of non-fastidious bacteria |
Each for $60.50
|
|
|||||
S04345
|
LaMotte™
5551 |
Environmental education: selective isolation of Fungi (Yeasts and molds) (SAB) |
Each for $60.50
|
|
|||||
S04346
|
LaMotte™
5552 |
Environmental education: cultivation of a wide variety of aerobic and anaerobic microorganisms (TSA), selective enumeration and cultivation of Yeasts, molds and Actinomycetes from food and other surfaces (RB) |
Each for $60.50
|
|
|||||
S04347
|
LaMotte™
5553 |
Environmental education: isolation and differentiation of gram (-) enteric bacilli, Coliform testing/recovery of stressed Coliforms |
Each for $60.50
|
|
|||||
Description
Flexible, ready-to-use paddles identify and quantify microbes in air, soil, water or on any surface.
- Longer shelf life than traditional Petri dishes
- No refrigeration required
- No need for inoculating loops or Bunsen burners
- Requires simple incubation and a magnifier
- Unique design enhances colony morphology characterization
- Ten paddles per box
Sabouraud Dextrose Agar: For selective cultivation of fungi (yeasts and molds).
Tryptic Soy (TSA)/Rose Bengal (RB) Agar: For cultivation of a wide variety of microorganisms (TSA) and selective isolation of yeasts and molds (RB).
Nutrient TTC/MacConkey: TTC: For field sampling cultivation and enumeration of coliform bacteria total coliform count; Gram (-) bacterial colonies appear as red dots; Gram (+) bacteria are usually inhibited. MAC: Medium gives improved differentiation between coliforms and nonlactose fermenting organisms; Gram (+) cocci are usually inhibited.
Order Info
Ships on Monday and Tuesday ONLY.
Specifications
18°C to 25°C | |
Sterile | |
Paddle |
United States | |
Free basic microbe identification application and access to technical information sheet | |
40 mL |
Safety and Handling
ShelfLife : 1 Year
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title