Learn More
3B Scientific™ Bionic Hybrid Auscultation Simulator (BHS)
When worn by the standardized patient, this suit simulates physiological conditions to test diagnostic and procedural skills. The Bionic Hybrid Auscultation Simulator is controlled wirelessly with easy to use software, responds in real time to diagnosis and treatment with direct feedback.
$14040.00
Specifications
For Use With (Application) | Testing Diagnostic and Procedural Skills |
---|---|
Includes | SimScope™ Wifi, Tablet, and Blood Pressure Cuff Accessory |
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
S27926
|
3B Scientific™
1022273 |
Medium |
Each for $14,040.00
|
|
|||||
S27927
|
3B Scientific™
1021981 |
Large |
Each for $14,040.00
|
|
|||||
S27928
|
3B Scientific™
1022275 |
X-Large |
Each for $14,040.00
|
|
|||||
S27929
|
3B Scientific™
1022278 |
2X-Large |
Each for $14,040.00
|
|
|||||
Description
- Simulates physiological conditions to test diagnostic and procedural skills
- Suit is controlled wirelessly with easy to use software
- Responds in real time to diagnosis and treatment with direct feedback
- Can be worn by students and/or manikins
- Integrated RFID Sensor Tags
- Carotid, and Radial Pulse
- Features 5-wire EKG connections, pulse points, blood pressure cuff accessory, as well as auscultation capabilities
- Instructors can easily adjust and adapt the suit scenarios to fit many conditions wirelessly by using the SimScope™ Wifi
- Includes SimScope™ Wifi, Tablet and Blood Pressure Cuff Accessory
- Reusable and washable at 30°C
Specifications
Testing Diagnostic and Procedural Skills | |
SimScope™ Wifi, Tablet, and Blood Pressure Cuff Accessory |