Learn More
Thermo Scientific™ BioMate™ 160 UV-Vis Spectrophotometer, PROMO
Perform automated measurements of nucleic acids, proteins and cell cultures with the Thermo Scientific™ BioMate™ 160 UV-Vis spectrophotometer.
Supplier: Thermo Scientific™ 8403010001PR22
Description
The BioMate UV-Vis Spectrophotometer provides automation for high-throughput options and room-light resistance, allowing lid-open operation for improved speed and convenience. Software includes pre-programmed life science methods such as Nucleic Acid A260; Protein A280 and A205; Protein Colorimetric assays and OD600.
A full line of accessories, including automated cell changers and cell holders, sipper, and fiber optic probes are designed to simplify sampling and accommodate high throughput measurements and temperature control.
Features:
- 7-inch high-resolution touchscreen user interface
- Pre-programmed life science methods
- Networking capable (Wi-Fi adaptor available)
- Room light resistance for open-lid operation
- Sipper accessory
- Removable, washable sample compartment liner with magnetic placement and hold-down
- Accommodates cells up to 100 mm pathlength
- (Optional) 8-position cell changer with switchable turret
- (Optional) 4-position cell changer (long path cells)
- (Optional) Peltier-thermostatted single cell holder (20 °C - 60 °C)
- (Optional) Accessory for test-tubes up to 25 mm diameter and 150 mm height
Designed for:
- Advanced teaching labs
- R&D departments
- Applications requiring high throughput or temperature control
Specifications
±0.002 A | |
Single USB-A | |
7 in. High Definition Color Touchscreen, Tiltable, 800 x 1280 pixels | |
BioDrop Duo | |
UV-Vis Spectrophotometer | |
(At 500 nm after 1 hour warmup) <0.0005 A/hr. | |
Touchscreen | |
(RMS at 500 nm 60 consecutive measurements) ≤0.00020 A at 0 A at 260 and 500 nm, ≤0.00030 A at 1 A at 260 and 500 nm, ≤0.00040 A at 2 A at 260 and 500 nm | |
±0.002 A at 0.5 A, ±0.004 A at 1.0 A, ±0.008 A at 2.0 A | |
−2 to 3.5 A | |
Snap-on printer available | |
< 1.0%T 198 nm (KCl) < 0.05%T at 220 nm (NaI), < 0.03%T at 340 nm (NaNO2) | |
±0.5 nm | |
< ±0.2 nm |
Accommodates (optional): 8-position cell changer, 4-position cell changer (long path cells), Peltier thermostatted cell holder (20 to 60°C), Sipper accessory, Fiber optic probe coupler | |
Dual Silicon Photodiodes | |
35.5 x 38.5 x 19.5 cm | |
2 nm | |
190 to 1100 nm | |
External AC/DC converter. Voltage and Frequency (Hz) selected automatically, 100/240 V, 50/60 Hz | |
> 5 years typical, 3 years guaranteed | |
Double Beam | |
−3 to 5 A | |
±0.001 A at 1 A measured at 1.0 A at 546 nm | |
Slow, medium and fast - up to 1600 nm/min. | |
2 years standard with 1 year extension upon registration of the instrument with Thermo Fisher Scientific within 6 months of purchase | |
0.2 nm, 0.5 nm, 1 nm, 2 nm, 5 nm | |
7.5 kg |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.