Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
United Scientific™ Biohazard Disposable Bags
![SDP](/content/dam/fishersci/glyphs/sdp.png)
![SDP](/content/dam/fishersci/glyphs/sdp.png)
![SCI_ED](/content/dam/fishersci/glyphs/scied.png)
Biohazard bags provide a safe method of disposal for biohazard waste.
Supplier: United Scientific™ UNU40107
Alert:
Prop 65
Description
- Bags have a clearly printed biohazard symbol and pass ASTM 1922 tear resistance and ASTM 1709 dart impact tests.
- Steam autoclavable for sterilization of waste before disposal.
- Polypropylene bags include black biohazard symbol and precautionary procedures printed in four languages (English, Spanish, French, German) for added safety.
- High temperature steam autoclavable to 135°C (275°F) Pass ASTM 1922 Tear Resistance and ASTM 1709 Dart Impact Tests 55 microns (2.2 mil) thickness
Specifications
Autoclavable | |
Red | |
Biohazard Materials | |
Non-sterile | |
16 in. |
9 L | |
Single-use | |
Polypropylene | |
24.5 in. |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Warning: Cancer and reproductive harm
Spot an opportunity for improvement?
Provide Content Correction